Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006145.2 | NP_006136.1 | 1 | 344 | ||
GenBank mRNA | AK296609.1 | - | 1 | 209 | ||
AK312624.1 | - | 1 | 205 | |||
BC002352.2 | - | 1 | 168 | 2821008 | ||
BC019827.1 | - | 1 | 158 | 4152608 | ||
CR536485.1 | - | 1 | 134 | |||
CR541677.1 | - | 1 | 134 | |||
D49547.1 | - | 1 | 174 | |||
DB454482.1 | - | 1 | 270 | |||
HY032238.1 | - | 1 | 345 | |||
X62421.1 | - | 1 | 179 |