Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d10c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178650.3 | NP_848765.2 | 10 | 1697 | ||
GenBank mRNA | AA217314.1 | - | 1 | 184 | ||
AK038366.1 | - | 2 | 837 | |||
AK040487.1 | - | 10 | 1698 | |||
AK075803.1 | - | 10 | 1684 | |||
AK088119.1 | - | 1692 | ||||
AK131177.1 | - | 9 | 1627 | |||
AK146426.1 | - | 10 | 1691 | |||
AK153836.1 | - | 10 | 1696 | |||
AK154444.1 | - | 9 | 2088 | |||
BC094901.1 | - | 9 | 1672 |