Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc30a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178651.3 | NP_848766.2 | 16 | 1667 | ||
GenBank mRNA | AF263460.1 | - | 14 | 1221 | ||
AK028885.1 | - | 16 | 1634 | |||
AK033990.1 | - | 692 | ||||
AK035541.1 | - | 16 | 1654 | |||
AK038559.1 | - | 15 | 1573 | |||
AK082853.1 | - | 16 | 1669 | |||
AK143277.1 | - | 16 | 1668 | |||
AK149549.1 | - | 8 | 4118 | |||
AY682914.1 | - | 17 | 1660 | |||
BC023833.1 | - | 8 | 771 | |||
BC024849.1 | - | 3 | 400 | |||
BC026565.1 | - | 12 | 1085 | |||
BC027806.1 | - | 16 | 1429 | |||
BC031705.1 | - | 13 | 1181 | |||
BC055773.1 | - | 13 | 1120 | |||
BC078440.1 | - | 14 | 1244 | |||
BC141008.1 | - | 17 | 1633 | 9055620 |