Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sccpdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178653.3 | NP_848768.1 | 7 | 840 | ||
GenBank mRNA | AK042919.1 | - | 7 | 728 | ||
AK046033.1 | - | 7 | 841 | |||
AK046468.1 | - | 7 | 828 | |||
AK050554.1 | - | 7 | 789 | |||
AK077695.1 | - | 7 | 790 | |||
AK082803.1 | - | 7 | 725 | |||
AK086448.1 | - | 7 | 863 | |||
AK086632.1 | - | 7 | 838 | |||
AK145399.1 | - | 7 | 824 | |||
AK166018.1 | - | 7 | 752 | |||
BC025803.1 | - | 7 | 837 | 5100552 |