Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sccpdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178653.3 | NP_848768.1 | 12 | 1460 | ||
GenBank mRNA | AK042919.1 | - | 12 | 1348 | ||
AK046033.1 | - | 12 | 1461 | |||
AK046468.1 | - | 12 | 1448 | |||
AK050554.1 | - | 12 | 1409 | |||
AK053698.1 | - | 1 | 2882 | |||
AK077695.1 | - | 12 | 1410 | |||
AK082803.1 | - | 12 | 1345 | |||
AK086448.1 | - | 12 | 1483 | |||
AK086632.1 | - | 12 | 1459 | |||
AK145399.1 | - | 9 | 1126 | |||
AK166018.1 | - | 12 | 1372 | |||
BC025803.1 | - | 12 | 1457 | 5100552 |