Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291999.1 | NP_001278928.1 | 2 | 317 | ||
NM_006153.5 | NP_006144.1 | 2 | 283 | |||
GenBank mRNA | AK312692.1 | - | 2 | 227 | ||
BC006403.2 | - | 2 | 260 | 4304621 | ||
DA265476.1 | - | 2 | 283 | |||
DA855124.1 | - | 2 | 317 | |||
DB117533.1 | - | 2 | 283 | |||
KU178218.1 | - | 1 | 153 | |||
KU178219.1 | - | 1 | 153 | |||
KU178220.1 | - | 1 | 153 | |||
X17576.1 | - | 2 | 254 |