Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Themis | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB509340.1 | - | 3, 4 | 793 | ||
AK040594.1 | - | 3, 4 | 972 | |||
AK041386.1 | - | 3, 4 | 1009 | |||
AK041562.1 | - | 3, 4 | 898 | |||
AK088136.1 | - | 3, 4 | 974 | |||
AK138701.1 | - | 3, 4 | 972 | |||
AK169558.1 | - | 4, 5 | 904 | |||
BC125486.1 | - | 3, 4 | 855 | 40130001 | ||
BC125488.1 | - | 3, 4 | 855 | 40130003 |