Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pld4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178911.4 | NP_849242.1 | 11 | 1629 | ||
GenBank mRNA | AK045459.1 | - | 11 | 1589 | ||
AK079549.1 | - | 11 | 1588 | |||
AK079878.1 | - | 11 | 1631 | |||
AK151551.1 | - | 11 | 1863 | |||
AK152969.1 | - | 10 | 1478 | |||
AK170362.1 | - | 11 | 1586 |