Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fancm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178912.3 | NP_849243.2 | 20 | 5010 | ||
GenBank mRNA | AK028941.1 | - | 3 | 379 | ||
AK173211.1 | - | 20 | 5030 | |||
BC150785.1 | - | 20 | 5004 | 9087696 |