Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fancm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178912.3 | NP_849243.2 | 23 | 6297 | ||
GenBank mRNA | AK028941.1 | - | 6 | 1667 | ||
AK050832.1 | - | 2 | 413 | |||
AK162428.1 | - | 4 | 1337 | |||
AK173211.1 | - | 23 | 6317 | |||
BC150785.1 | - | 23 | 6291 | 9087696 |