Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spata7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289573.1 | NP_001276502.1 | 3 | 332 | ||
NM_178914.4 | NP_849245.2 | 3 | 332 | |||
GenBank mRNA | AK036359.1 | - | 3 | 317 | ||
AK054041.1 | - | 1 | 1277 | |||
AK077500.1 | - | 3 | 243 | |||
AK153698.1 | - | 3 | 308 | |||
AK169473.1 | - | 3 | 313 | |||
BC046960.1 | - | 3 | 290 | 6391450 | ||
BY252962.1 | - | 3 | 331 |