Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spata7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289572.1 | NP_001276501.1 | 5 | 874 | ||
NM_001289574.1 | NP_001276503.1 | 6 | 1036 | |||
NM_178914.4 | NP_849245.2 | 6 | 970 | |||
GenBank mRNA | AK036359.1 | - | 6 | 954 | ||
AK077500.1 | - | 6 | 881 | |||
AK161308.1 | - | 5 | 820 | |||
BC046960.1 | - | 6 | 928 | 6391450 | ||
DQ196101.1 | - | 5 | 700 |