Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem179 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178915.3 | NP_849246.2 | 4 | 1833 | ||
GenBank mRNA | AK044254.1 | - | 4 | 1810 | ||
AK047810.1 | - | 4 | 1849 | |||
AK049973.1 | - | 4 | 1801 | |||
AK090230.1 | - | 4 | 1850 | |||
BC046482.1 | - | 4 | 1842 | 5357484 | ||
BC055014.1 | - | 4 | 1825 | 6493726 |