Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kank1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181404.5 | NP_852069.4 | 14 | 5246 | ||
GenBank mRNA | AK084638.1 | - | 7 | 1696 | ||
AK122214.1 | - | 10 | 3612 | |||
BC037712.1 | - | 6 | 1579 | |||
BC050250.1 | - | 9 | 2053 | |||
BC058673.1 | - | 10 | 2784 | |||
BC079563.1 | - | 10 | 4573 | 30535512 |