Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Me3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181407.4 | NP_852072.2 | 3 | 458 | ||
GenBank mRNA | AK032196.1 | - | 3 | 459 | ||
AK035780.1 | - | 2 | 79 | |||
AK080798.1 | - | 3 | 391 | |||
AK133593.1 | - | 4 | 446 | |||
AK209005.1 | - | 2 | 144 |