Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tm6sf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293795.1 | NP_001280724.1 | 3 | 313 | ||
NM_181540.4 | NP_853518.1 | 3 | 313 | |||
GenBank mRNA | AK077126.1 | - | 3 | 306 | ||
BB869951.1 | - | 3 | 314 | |||
BC024498.1 | - | 3 | 301 | 4982620 |