Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181849.2 | NP_862897.1 | 6 | 911 | ||
GenBank mRNA | AF413205.1 | - | 2 | 183 | ||
AK011118.1 | - | 6 | 930 | |||
AK011920.1 | - | 6 | 926 | |||
AK146960.1 | - | 6 | 931 | |||
AK168626.1 | - | 6 | 925 | |||
BC031715.1 | - | 6 | 911 | 4195310 |