Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122780.1 | NP_001116252.1 | 2 | 944 | ||
NM_029652.1 | NP_083928.1 | 2 | 955 | |||
NM_182782.2 | NP_877583.1 | 2 | 955 | |||
GenBank mRNA | AK143086.1 | - | 2 | 901 | ||
BC011221.1 | - | 2 | 886 | |||
BC022930.1 | - | 2 | 886 | |||
BC027373.1 | - | 2 | 957 | 4948451 |