Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182782.2 | NP_877583.1 | 2 | 2074 | ||
GenBank mRNA | AK157879.1 | - | 2 | 1123 | ||
BC027373.1 | - | 2 | 2076 | 4948451 |