Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163263.1 | NP_001156735.1 | 2, 3 | 621 | ||
NM_182999.2 | NP_892044.1 | 3, 4 | 468 | |||
GenBank mRNA | AK048862.1 | - | 2, 3 | 580 | ||
AK051278.1 | - | 2, 3 | 580 | |||
AK053783.1 | - | 3, 4 | 456 | |||
AK082529.1 | - | 2, 3 | 524 | |||
AK085436.1 | - | 2, 3 | 599 | |||
AK087860.1 | - | 2, 3 | 545 | |||
AK139883.1 | - | 3, 4 | 446 | |||
AK220492.1 | - | 2, 3 | 537 | |||
BC052482.1 | - | 3, 4 | 407 | 6308280 | ||
CJ052397.1 | - | 3, 4 | 449 |