Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slain1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198014.2 | NP_932131.2 | 7 | 1995 | ||
GenBank mRNA | AK036190.1 | - | 9 | 2100 | ||
BC025223.1 | - | 5 | 1149 | 5010865 | ||
BC079866.1 | - | 7 | 1995 | 6811096 |