Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam175b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198017.3 | NP_932134.3 | 6 | 515 | ||
GenBank mRNA | AK082887.1 | - | 6 | 496 | ||
AK146064.1 | - | 6 | 490 | |||
AK163824.1 | - | 1 | 1559 | |||
AK170704.1 | - | 6 | 490 | |||
AK171524.1 | - | 6 | 441 | |||
BC030838.1 | - | 3 | 271 |