Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpeb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290826.1 | NP_001277755.1 | 11 | 3330 | ||
NM_001290827.1 | NP_001277756.1 | 11 | 2979 | |||
NM_001290828.1 | NP_001277757.1 | 11 | 2972 | |||
NM_001290829.1 | NP_001277758.1 | 10 | 2829 | |||
NM_198300.3 | NP_938042.2 | 11 | 3041 | |||
GenBank mRNA | AB093274.1 | - | 11 | 2970 | ||
AK044639.1 | - | 11 | 2974 | |||
AK147243.1 | - | 11 | 3331 | |||
AY313774.1 | - | 11 | 2863 |