Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnck | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199351.1 | NP_001186280.1 | 10, 11 | 1092 | ||
NM_001199352.1 | NP_001186281.1 | 10, 11 | 956 | |||
NM_012040.3 | NP_036170.1 | 10, 11 | 1032 | |||
GenBank mRNA | AB023027.1 | - | 9, 10 | 902 | ||
AF181984.1 | - | 10, 11 | 1006 | |||
AK134665.1 | - | 10, 11 | 963 | |||
AK158755.1 | - | 10, 11 | 957 | |||
BC051996.1 | - | 10, 11 | 939 | |||
BC055891.1 | - | 10, 11 | 932 | 4208888 |