Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agpat6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018743.4 | NP_061213.2 | 13 | 2289 | ||
GenBank mRNA | AF015811.1 | - | 10 | 964 | ||
AF406611.1 | - | 12 | 1800 | |||
AK045235.1 | - | 13 | 2183 | |||
AK077561.1 | - | 13 | 2173 | |||
AK083589.1 | - | 13 | 2183 | |||
AK161270.1 | - | 13 | 2152 | |||
AK162033.1 | - | 10 | 1094 | |||
AK167876.1 | - | 13 | 2197 | |||
AK169257.1 | - | 13 | 2183 | |||
AY489184.1 | - | 12 | 1362 | |||
BC031767.1 | - | 12 | 1643 | 4457846 | ||
BC057860.1 | - | 10 | 963 |