Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252580.1 | NP_001239509.1 | 2 | 305 | ||
NM_019729.3 | NP_062703.2 | 2 | 305 | |||
GenBank mRNA | AB045709.1 | - | 1 | 79 | ||
AF057146.1 | - | 1 | 96 | |||
AK040791.1 | - | 2 | 252 | |||
AK122195.1 | - | 2 | 216 | |||
AK129003.1 | - | 2 | 273 | |||
AK134058.1 | - | 2 | 260 | |||
AK153015.1 | - | 2 | 216 | |||
AK163469.1 | - | 2 | 238 | |||
AK166540.1 | - | 2 | 306 | |||
BC038123.1 | - | 2 | 290 | |||
BC050947.1 | - | 2 | 270 | 6825981 | ||
BC061465.1 | - | 2 | 270 | 6825981 | ||
BC066126.1 | - | 2 | 268 | 30301665 | ||
BY147904.1 | - | 2 | 305 |