Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019729.3 | NP_062703.2 | 2, 3 | 329 | ||
GenBank mRNA | AB045709.1 | - | 1, 2 | 103 | ||
AF057146.1 | - | 1, 2 | 120 | |||
AK040791.1 | - | 2, 3 | 276 | |||
AK122195.1 | - | 2, 3 | 240 | |||
AK134058.1 | - | 2, 3 | 284 | |||
AK153015.1 | - | 2, 3 | 240 | |||
AK163469.1 | - | 2, 3 | 262 | |||
AK166540.1 | - | 2, 3 | 330 | |||
BC038123.1 | - | 2, 3 | 314 | |||
BC050947.1 | - | 2, 3 | 294 | 6825981 | ||
BC061465.1 | - | 2, 3 | 294 | 6825981 | ||
BC066126.1 | - | 2, 3 | 292 | 30301665 |