Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sirt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019812.2 | NP_062786.1 | 2 | 481 | ||
GenBank mRNA | AF214646.1 | - | 2 | 466 | ||
AK140330.1 | - | 2 | 429 | |||
AY377984.1 | - | 2 | 466 | |||
BY740019.1 | - | 2 | 482 |