Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Herpud2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020586.2 | NP_065611.1 | 3, 4 | 1152 | ||
GenBank mRNA | AB041580.1 | - | 3, 4 | 947 | ||
AK019862.1 | - | 3, 4 | 1097 | |||
AK135935.1 | - | 3, 4 | 1153 | |||
BC029691.1 | - | 3, 4 | 370 | 5375603 | ||
BC043693.1 | - | 3, 4 | 1191 | 5150485 |