Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Qsox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024945.1 | NP_001020116.1 | 6 | 767 | ||
NM_023268.2 | NP_075757.1 | 6 | 767 | |||
GenBank mRNA | AB044284.1 | - | 6 | 675 | ||
AK004880.1 | - | 6 | 713 | |||
AK083938.1 | - | 6 | 718 | |||
AK166839.1 | - | 6 | 713 | |||
AK169877.1 | - | 6 | 720 | |||
AK169920.1 | - | 6 | 716 | |||
AK170449.1 | - | 6 | 680 | |||
BC034131.1 | - | 3 | 259 | |||
BC076590.1 | - | 6 | 721 | 30548238 |