Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tinagl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168333.1 | NP_001161805.1 | 9, 10 | 1186 | ||
NM_023476.3 | NP_075965.2 | 9, 10 | 1327 | |||
GenBank mRNA | AB047402.1 | - | 9, 10 | 1328 | ||
AB050626.1 | - | 9, 10 | 1164 | |||
AK087142.1 | - | 1187 | ||||
BC005738.1 | - | 9, 10 | 1154 | 2650851 | ||
BC018539.1 | - | 9, 10 | 1171 | 3596560 | ||
CT010275.1 | - | 8, 9 | 945 |