Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tinagl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168333.1 | NP_001161805.1 | 12 | 1726 | ||
NM_023476.3 | NP_075965.2 | 12 | 1867 | |||
GenBank mRNA | AB047402.1 | - | 12 | 1868 | ||
AB050626.1 | - | 12 | 1704 | |||
AK003901.1 | - | 3 | 517 | |||
AK219988.1 | - | 1 | 42 | |||
BC005738.1 | - | 12 | 1694 | 2650851 | ||
BC018539.1 | - | 12 | 1711 | 3596560 |