Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nipa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256130.1 | NP_001243059.1 | 4 | 661 | ||
NM_001256131.1 | NP_001243060.1 | 3 | 540 | |||
NM_001256132.1 | NP_001243061.1 | 4 | 465 | |||
NM_001256133.1 | NP_001243062.1 | 2 | 115 | |||
NM_023647.6 | NP_076136.2 | 4 | 1415 | |||
GenBank mRNA | AB041581.1 | - | 4 | 598 | ||
AK008106.1 | - | 4 | 648 | |||
AK011248.1 | - | 3 | 487 | |||
AK012859.1 | - | 4 | 599 | |||
AK028734.1 | - | 4 | 1373 | |||
AK031255.1 | - | 4 | 626 | |||
AK041427.1 | - | 4 | 662 | |||
AK076051.1 | - | 4 | 618 | |||
AK076184.1 | - | 4 | 602 | |||
AK154755.1 | - | 4 | 619 | |||
AK183552.1 | - | 3 | 372 | |||
AK196701.1 | - | 4 | 387 | |||
AV495155.1 | - | 2 | 58 | |||
BC031109.1 | - | 1 | 1929 | |||
BC038499.1 | - | 4 | 603 | 4482150 |