Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nipa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256130.1 | NP_001243059.1 | 7 | 1104 | ||
NM_001256131.1 | NP_001243060.1 | 6 | 983 | |||
NM_001256132.1 | NP_001243061.1 | 7 | 908 | |||
NM_001256133.1 | NP_001243062.1 | 5 | 558 | |||
NM_023647.6 | NP_076136.2 | 7 | 1858 | |||
GenBank mRNA | AB041581.1 | - | 7 | 1041 | ||
AK008106.1 | - | 7 | 1091 | |||
AK011248.1 | - | 6 | 928 | |||
AK012859.1 | - | 7 | 1042 | |||
AK028734.1 | - | 7 | 1816 | |||
AK041427.1 | - | 7 | 1105 | |||
AK076051.1 | - | 7 | 1061 | |||
AK076184.1 | - | 7 | 1045 | |||
AK154755.1 | - | 7 | 1062 | |||
BC038499.1 | - | 7 | 1046 | 4482150 |