Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nipa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256130.1 | NP_001243059.1 | 6 | 926 | ||
NM_001256131.1 | NP_001243060.1 | 5 | 805 | |||
NM_001256132.1 | NP_001243061.1 | 6 | 730 | |||
NM_001256133.1 | NP_001243062.1 | 4 | 380 | |||
NM_023647.6 | NP_076136.2 | 6 | 1680 | |||
GenBank mRNA | AB041581.1 | - | 6 | 863 | ||
AK008106.1 | - | 6 | 913 | |||
AK011248.1 | - | 5 | 750 | |||
AK012859.1 | - | 6 | 864 | |||
AK028734.1 | - | 6 | 1638 | |||
AK041427.1 | - | 6 | 927 | |||
AK076051.1 | - | 6 | 883 | |||
AK076184.1 | - | 6 | 867 | |||
AK154755.1 | - | 6 | 884 | |||
BC038499.1 | - | 6 | 868 | 4482150 |