Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trp53rk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023815.4 | NP_076304.2 | 2 | 1370 | ||
GenBank mRNA | AK028411.1 | - | 2 | 1371 | ||
AK046261.1 | - | 2 | 1348 | |||
AK049291.1 | - | 2 | 1355 | |||
BC017155.1 | - | 2 | 1388 | 4910610 |