Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ralgps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159965.1 | NP_001153437.1 | 6 | 531 | ||
NM_001159966.1 | NP_001153438.1 | 6 | 429 | |||
NM_001159967.1 | NP_001153439.1 | 6 | 531 | |||
NM_001159968.1 | NP_001153440.1 | 5 | 426 | |||
NM_023884.4 | NP_076373.3 | 6 | 471 | |||
GenBank mRNA | AF312924.1 | - | 5 | 379 | ||
AK018622.1 | - | 6 | 473 | |||
AK019543.1 | - | 5 | 429 | |||
AK029049.1 | - | 6 | 472 | |||
AK029993.1 | - | 7 | 609 | |||
AK033549.1 | - | 6 | 427 | |||
AK036803.1 | - | 5 | 403 | |||
AK161080.1 | - | 6 | 498 | |||
AK165002.1 | - | 6 | 423 | |||
AK169368.1 | - | 6 | 449 | |||
BC043132.1 | - | 6 | 436 | 6811441 | ||
BC052663.1 | - | 6 | 448 | 30051209 |