Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Icam4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023892.2 | NP_076381.1 | 2 | 736 | ||
GenBank mRNA | AF296282.1 | - | 2 | 655 | ||
AF296283.1 | - | 2 | 655 | |||
AK019013.1 | - | 2 | 438 | |||
BC116909.1 | - | 2 | 736 | 40126228 | ||
BC116913.1 | - | 2 | 736 | 40126232 |