Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsc22d4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023910.6 | NP_076399.4 | 5 | 2020 | ||
GenBank mRNA | AF201286.1 | - | 5 | 1733 | ||
AK018735.2 | - | 5 | 1993 | |||
AK040534.1 | - | 5 | 1957 | |||
AK143608.1 | - | 5 | 1797 | |||
AK150043.1 | - | 5 | 1989 | |||
AK154153.1 | - | 5 | 1970 | |||
AK154490.1 | - | 5 | 1620 | |||
BC018544.1 | - | 4 | 1492 | 3666221 | ||
BC023761.1 | - | 5 | 1924 | 5343927 |