Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdac9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271386.1 | NP_001258315.1 | 5 | 490 | ||
NM_024124.3 | NP_077038.2 | 3 | 549 | |||
GenBank mRNA | AF235053.1 | - | 2 | 218 | ||
AF279371.1 | - | 2 | 218 | |||
AF324492.1 | - | 2 | 218 | |||
AK087563.1 | - | 5 | 839 | |||
AK133334.1 | - | 5 | 490 | |||
BC098187.1 | - | 3 | 534 | 5363624 |