Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdac9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024124.3 | NP_077038.2 | 12 | 2055 | ||
GenBank mRNA | AF235053.1 | - | 11 | 1724 | ||
AF279371.1 | - | 10 | 1592 | |||
AF324492.1 | - | 11 | 1718 | |||
AK144242.1 | - | 3 | 530 | |||
BC098187.1 | - | 12 | 2040 | 5363624 |