Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bhlhe41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271768.1 | NP_001258697.1 | 3 | 359 | ||
NM_024469.2 | NP_077789.1 | 3 | 359 | |||
GenBank mRNA | AA013582.1 | - | 3 | 360 | ||
AB044090.1 | - | 3 | 279 | |||
BC023231.1 | - | 3 | 311 | |||
BC037742.1 | - | 3 | 319 | |||
BC051555.1 | - | 3 | 320 |