Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136528.1 | NP_001130000.1 | 2 | 362 | ||
NM_001136530.1 | NP_001130002.1 | 2 | 292 | |||
NM_006216.3 | NP_006207.1 | 2 | 362 | |||
NR_073116.1 | 3 | 780 | ||||
XM_005246641.2 | XP_005246698.1 | 2 | 451 | |||
XM_017004329.1 | XP_016859818.1 | 2 | 343 | |||
XM_017004330.1 | XP_016859819.1 | 2 | 180 | |||
XM_017004331.1 | XP_016859820.1 | 2 | 343 | |||
XM_017004332.1 | XP_016859821.1 | 2 | 180 | |||
GenBank mRNA | A03911.1 | - | 1 | 143 | ||
AK225823.1 | - | 2 | 331 | |||
AK295564.1 | - | 2 | 260 | |||
AK297588.1 | - | 2 | 167 | |||
AK312499.1 | - | 2 | 376 | |||
BC015663.2 | - | 2 | 319 | 4578406 | ||
BC042628.1 | - | 3 | 779 | 4824856 | ||
DC304261.1 | - | 2 | 292 | |||
DC366668.1 | - | 2 | 363 | |||
M17783.1 | - | 1 | 119 |