Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ublcp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024475.5 | NP_077795.2 | 10 | 992 | ||
GenBank mRNA | AK043952.1 | - | 10 | 899 | ||
AK050031.1 | - | 10 | 903 | |||
AK147744.1 | - | 10 | 940 | |||
AK153998.1 | - | 10 | 904 | |||
BC002236.1 | - | 3 | 266 | |||
BC056652.1 | - | 6 | 493 | |||
BC083331.1 | - | 8 | 653 | |||
BC085111.1 | - | 9 | 895 | 30735741 | ||
BC094051.1 | - | 4 | 298 | |||
BC106093.1 | - | 9 | 869 | 5098953 |