Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3bp5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK086088.1 | - | 7 | 2633 | ||
AK122543.1 | - | 5 | 5836 | |||
AK144928.1 | - | 7 | 2631 | |||
AK150129.1 | - | 7 | 2611 | |||
AK156171.1 | - | 7 | 2672 | |||
AK159202.1 | - | 7 | 2663 | |||
AK166491.1 | - | 7 | 2632 | |||
AK167203.1 | - | 7 | 2749 | |||
AK172333.1 | - | 7 | 2672 | |||
AK206009.1 | - | 1 | 42 | |||
AK208174.1 | - | 1 | 33 | |||
BC003251.1 | - | 6 | 2293 | 3500047 | ||
BC013661.1 | - | 3 | 1764 | |||
BC094418.1 | - | 7 | 2654 | 30702568 |