Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mien1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025559.2 | NP_079835.1 | 4 | 646 | ||
GenBank mRNA | AK002959.1 | - | 4 | 629 | ||
AK007795.1 | - | 4 | 629 | |||
AK009922.1 | - | 4 | 624 | |||
AK041314.1 | - | 4 | 628 | |||
BC021589.1 | - | 4 | 630 | 5100542 | ||
CA530220.1 | - | 3 | 472 |