Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtmr14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026849.2 | NP_081125.2 | 18 | 1877 | ||
GenBank mRNA | AK034867.1 | - | 17 | 1738 | ||
BC012436.1 | - | 16 | 1405 | |||
BC018294.1 | - | 18 | 1827 | 4021243 | ||
DV653632.1 | - | 6 | 621 |