Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ints4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027256.2 | NP_081532.1 | 4, 5 | 623 | ||
GenBank mRNA | AK088635.1 | - | 4, 5 | 567 | ||
AK163383.1 | - | 4, 5 | 577 | |||
BC013710.1 | - | 4, 5 | 569 | 3156446 | ||
CA450656.1 | - | 4, 5 | 493 | |||
CX565861.1 | - | 1, 2 | 107 |