Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLTP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242920.1 | NP_001229849.1 | 3 | 480 | ||
NM_006227.3 | NP_006218.1 | 3 | 480 | |||
NM_182676.2 | NP_872617.1 | 3 | 480 | |||
GenBank mRNA | AB076694.1 | - | 3 | 264 | ||
AK092397.1 | - | 3 | 221 | |||
AK092523.1 | - | 3 | 268 | |||
AK097009.1 | - | 2 | 778 | |||
AK222690.1 | - | 3 | 268 | |||
AK289371.1 | - | 3 | 268 | |||
AK299181.1 | - | 3 | 272 | |||
BC005045.1 | - | 3 | 261 | 3510757 | ||
BC019847.1 | - | 3 | 262 | 4997011 | ||
BC019898.1 | - | 3 | 255 | 4992839 | ||
L26232.1 | - | 3 | 270 |