Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127218.2 | NP_001120690.1 | 4 | 733 | ||
NM_001256879.1 | NP_001243808.1 | 5 | 1094 | |||
NM_006230.3 | NP_006221.2 | 4 | 560 | |||
XM_011515431.1 | XP_011513733.1 | 4 | 556 | |||
GenBank mRNA | AK292347.1 | - | 5 | 1094 | ||
AK298168.1 | - | 4 | 424 | |||
AK312294.1 | - | 4 | 693 | |||
AU117477.2 | - | 4 | 538 | |||
BC000459.2 | - | 4 | 523 | 2822169 | ||
BC030001.2 | - | 3 | 500 | |||
U21090.1 | - | 4 | 522 |